missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Magnesium rod, 6.35mm (0.25 in.) dia. x 25.4mm (1.0in) length, 99.95% (metals basis)

Manufacturer:  Alfa Aesar™ 47073KJ

 View more versions of this product

Catalog No. AA47073KJ



Specifications

Specifications

Magnesium rod
5 pc.
7439-95-4
Mg
magnesio, sheet, powdered, magnesio italian, turnings, rieke's active, ribbon, unii-i38zp9992a, hsdb 654, nitrate, acs
[Mg]
Odorless
5462224
Pieces
≈1.4g/piece
6.35mm (0.25 in.) dia. x 25.4mm (1.0 in.) length
(metals basis)
MFCD00085308
FYYHWMGAXLPEAU-UHFFFAOYSA-N
magnesium
24.305
CHEBI:25107
99.95%

Chemical Identifiers

7439-95-4
24.305
FYYHWMGAXLPEAU-UHFFFAOYSA-N
5462224
magnesium
Mg
MFCD00085308
magnesio, sheet, powdered, magnesio italian, turnings, rieke's active, ribbon, unii-i38zp9992a, hsdb 654, nitrate, acs
CHEBI:25107
[Mg]
Safety and Handling

Safety and Handling

RTECSNumber : OM2100000

TSCA : TSCA

SDS Documents