missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Magnesium rod, 3.3cm (1.3 in.) dia. x 30cm (12 in.) long, 99.8% (metals basis)

Manufacturer:  Alfa Aesar™ 10231KM

 View more versions of this product

Catalog No. AA10231KM



Specifications

Specifications

Magnesium rod
25 pc.
7439-95-4
Mg
magnesio, sheet, powdered, magnesio italian, turnings, rieke's active, ribbon, unii-i38zp9992a, hsdb 654, nitrate, acs
[Mg]
Odorless
5462224
Rod
≈450g/pc
3.3cm (1.3 in.) dia. x 30cm (12 in.) long
(metals basis)
MFCD00085308
FYYHWMGAXLPEAU-UHFFFAOYSA-N
magnesium
24.305
CHEBI:25107
99.8%

Chemical Identifiers

7439-95-4
24.305
FYYHWMGAXLPEAU-UHFFFAOYSA-N
5462224
magnesium
Mg
MFCD00085308
magnesio, sheet, powdered, magnesio italian, turnings, rieke's active, ribbon, unii-i38zp9992a, hsdb 654, nitrate, acs
CHEBI:25107
[Mg]
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents