missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Magnesium ribbon, approximate dimensions, 3.0mm (0.12in) wide, 0.15mm (0.0059 in.) thick, Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific 1281114

 View more versions of this product

Catalog No. AA1281114



Specifications

Specifications

Magnesium ribbon
25g
7439-95-4
MFCD00085308
hsdb 654
[Mg]
Odorless
5462224
≈0.78g/m
≈0.78g/m
Approximate dimensions: 3.0mm (0.12 in.) wide, 0.15mm (0.0059 in.) thick
Mg
UN1869
FYYHWMGAXLPEAU-UHFFFAOYSA-N
magnesium
24.305
CHEBI:25107

Chemical Identifiers

7439-95-4
24.305
FYYHWMGAXLPEAU-UHFFFAOYSA-N
5462224
magnesium
Mg
MFCD00085308
hsdb 654
CHEBI:25107
[Mg]
Safety and Handling

Safety and Handling

GHS H Statement
H228-H251-H261
Flammable solid.
Self-heating: may catch fire.
In contact with water releases flammable gas.

P210-P223-P231+P232-P235+P410-P240-P241-P280-P335+P334-P370+P378s-P501c

H228-H252-H261

DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: MAGNESIUM

EINECSNumber : 231-104-6

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents