missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Magnesium, Reagent, Ribbon, +99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 413380250

 View more versions of this product

Catalog No. AC413380250


 

Specifications Description & Specifications

Specifications

≥99.0%
Metal Ribbon
Silvery white
651.0°C
500°C
01,627; 02,254; 03,189; 04,315; 05,419; 06,351; 07,218; 10,251; 12,290; 13,170; 15,120; 16,198
50ppm max.
Mg
Magnesium
Mg
200ppm max.
500ppm max.
24.3
25g
1869
1107.0°C
1.74
15, 5717
50ppm max.
MFCD00085308
99+%
50ppm max.
99+%
7439-95-4

Chemical Identifiers

7439-95-4
99+%
Mg
MFCD00085308
≥99.0%
Metal Ribbon
Mg
24.3
Magnesium, 99+%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
In contact with water releases flammable gases.
Self-heating in large quantities; may catch fire.
Flammable solid.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Keep away from any possible contact with water, because of violent reaction and possible flash fire.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 231-104-6

TSCA : TSCA