Please login to your online account to display your discounted pricing

Magnesium, Reagent, Ribbon, +99%, ACROS Organics™


Quantity 100g

Manufacturer: thermo fisher scientific  AC413381000

 View more versions of this product

Catalog No. AC413381000

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Metal Ribbon
Assay ≥99.0%
Formula Weight 24.3
Quantity 100g
Color Silvery white
Merck Index 15, 5717
UN Number 1869
Melting Point 651.0°C
Boiling Point 1107.0°C
Flash Point 500°C
EINECS Number 231-104-6
TSCA TSCA
Density 1.74
Fieser 01,627; 02,254; 03,189; 04,315; 05,419; 06,351; 07,218; 10,251; 12,290; 13,170; 15,120; 16,198
Chemical Name or Material Magnesium
Copper (Cu) 50ppm max.
Iron (Fe) 500ppm max.
Linear Formula Mg
Nickel (Ni) 50ppm max.
Health Hazard 2 GHS H Statement:
In contact with water releases flammable gases.
Self-heating in large quantities; may catch fire.
Flammable solid.
Health Hazard 3 GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Keep away from any possible contact with water, because of violent reaction and possible flash fire.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Zinc (Zn) 200ppm max.
Name Note 99+%
Lead (Pb) 50ppm max.
Assay Percent Range 99+%
Molecular Formula Mg
MDL Number MFCD00085308
CAS 7439-95-4
Health Hazard 1 Danger

Specifications

Chemical Name or Material Magnesium
Name Note 99+%
CAS 7439-95-4
Physical Form Metal Ribbon
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula Mg
Linear Formula Mg
Formula Weight 24.3
MDL Number MFCD00085308
Accessories

Danger

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Keep away from any possible contact with water, because of violent reaction and possible flash fire.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
In contact with water releases flammable gases.
Self-heating in large quantities; may catch fire.
Flammable solid.

SDS & Safety Documents