Please login to your online account to display your discounted pricing

Magnesium Reagent, LabChem™

$47.23 - $47.23

Chemical Identifiers

Chemical Name or Material Magnesium Reagent
CAS 7732-18-5
CAS Max % 98.5
CAS 1310-73-2
CAS Max % 1
CAS 74-39-5
CAS Max % 0.5
Linear Formula C12H9N3O4
Formula Weight 259.22
UN Number UN1824
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC164707

 
lab chem
LC164707
125mL Each for $47.23
Description & Specifications

Specifications

Formula Weight 259.22
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
Causes severe skin burns and eye damage.
CAS 1310-73-2
CAS Max % 98.5
Health Hazard 3 GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
CAS 74-39-5
Color Clear, red-purple
Packaging Poly Bottle
CAS Max % 0.5
Quantity 125mL
UN Number UN1824
Linear Formula C12H9N3O4
Chemical Name or Material Magnesium Reagent
Physical Form Liquid
Decomposition Information Nitrogen oxides; Carbon monoxide; Carbon dioxide
CAS Max % 1
Solubility Soluble in water
Identification Passes Test
Storage Information Room Temperature
CAS 7732-18-5
Safety and Handling

Danger

GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Causes severe skin burns and eye damage.