Please login to your online account to display your discounted pricing

Magnesium powder, -325 mesh, 99.8%, Alfa Aesar

$77.59 - $77.59

Chemical Identifiers

Chemical Name or Material Magnesium
CAS 7439-95-4
Physical Form -325 Mesh Powder
Assay Percent Range 99.8%
Molecular Formula Mg
Formula Weight 24.3
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1023309

 
alfa aesar
1023309
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1023336

 
alfa aesar
1023336
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1023357

 
alfa aesar
1023357
4 x 500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1023322

 
alfa aesar
1023322
100g Each for $77.59
Description & Specifications

Specifications

Formula Weight 24.3
Quantity 10g
Physical Form Powder, -325 mesh
Molecular Formula Mg
Chemical Name or Material Magnesium powder
EINECS Number 231-104-6
Assay Percent Range 99.8%
MDL Number MFCD00085308
CAS 7439-95-4