Learn More
Magnesium, plasma standard solution, Specpure™ Mg 1000μg/mL, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 013861AE
Description
Magnesium, plasma standard solution is used as a standard solution in analytical chemistry and atomic absorption spectroscopy. It is also used as a single-element standard solution for plasma emission spectrometry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Magnesium | |
100 mL | |
Mg | |
JLVVSXFLKOJNIY-UHFFFAOYSA-N | |
magnesium | |
5462224 | |
Nitric acid: 5.0%; Magnesium: 0.1%; Water: 94.9% | |
Miscible with water |
7439-95-4,7697-37-2,7732-18-5 | |
Colorless | |
MFCD00085308 | |
[Mg] | |
24.30 | |
CHEBI:25107 | |
23 hPa (17mm Hg) at 20°C | |
Liquid |
Chemical Identifiers
7439-95-4 | |
24.30 | |
JLVVSXFLKOJNIY-UHFFFAOYSA-N | |
CHEBI:25107 | |
[Mg] |
Mg | |
MFCD00085308 | |
5462224 | |
magnesium |
Safety and Handling
P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
H290-H314-H335
DOTInformation : Hazard Class: 8, Packing Group: III
EINECSNumber : 231-714-2
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only