missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Magnesium perchlorate hexahydrate, 99% (metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 11635A1

 View more versions of this product

Catalog No. AA11635A1



Specifications

Specifications

Magnesium perchlorate hexahydrate
99% (Metals basis)
Cl2H12MgO14
UN1475
2clo4.mg.6h2o
O.O.O.O.O.O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Mg+2]
331.287
331.33 (223.23 Anhydrous)
99%
Hygroscopic
1kg
13446-19-0
(metals basis)
MFCD00149778
14,5678
MGBDNCBRZIMSDZ-UHFFFAOYSA-L
magnesium;diperchlorate;hexahydrate
16211969
Crystalline
Odorless
185°C to 190°C

Chemical Identifiers

13446-19-0
331.287
MGBDNCBRZIMSDZ-UHFFFAOYSA-L
16211969
O.O.O.O.O.O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Mg+2]
Cl2H12MgO14
MFCD00149778
2clo4.mg.6h2o
magnesium;diperchlorate;hexahydrate
Safety and Handling

Safety and Handling

GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P221-P210-P220-P305+P351+P338-P405-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Keep/Store away from clothing/combustible materials.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: MAGNESIUM PERCHLORATE

TSCA : TSCA

Documents