Learn More
Magnesium oxide substrate, 10x10x0.5mm, polished one side, 100 orientation, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 036427KT
Description
A useful laboratory reagent, and a good reference white color for colorimetry. The interaction of magnesium oxide substrates with nickel ferrite films has been investigated using a wide range of electron optical techniques. Considerable interdiffusion is observed, which is likely to alter the magnetic properties of these films. Epitaxial ferrite-spinel layer is formed on magnesium oxide substrate. Additionally, Magnesium oxide in nanoparticles dispersed in alcohol have been reported to inhibit hydrolytic and oxidative degradation of paper documents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Magnesium oxide substrate | |
1309-48-4 | |
MgO | |
magnesium oxide, magnesia, seawater magnesia, causmag, granmag, maglite, periclase, seasorb, animag | |
CPLXHLVBOLITMK-UHFFFAOYSA-N | |
oxomagnesium | |
14792 |
10x10x0.5mm, polished one side, 100 orientation | |
1 Ea. | |
MFCD00011109 | |
Soluble in 5 M HCl (0.1 M) at 20°C,and dilute acids. Insoluble in water,and ethanol. | |
O=[Mg] | |
40.30 |
Chemical Identifiers
1309-48-4 | |
40.30 | |
CPLXHLVBOLITMK-UHFFFAOYSA-N | |
14792 | |
O=[Mg] |
MgO | |
MFCD00011109 | |
magnesium oxide, magnesia, seawater magnesia, causmag, granmag, maglite, periclase, seasorb, animag | |
oxomagnesium |
Safety and Handling
EINECSNumber : 215-171-9
RTECSNumber : OM3850000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only