Please login to your online account to display your discounted pricing

Magnesium Oxide Substrate, 10 x 10 x 1mm, Polished One Side, 110 Orientation, Alfa Aesar™

100 Orientation

$132.07 - $594.16

Chemical Identifiers

Chemical Name or Material Magnesium Oxide Substrate
Name Note Polished Two Sides
Physical Form Plate
Molecular Formula MgO
MDL Number MFCD00011109
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA40274KT

 
alfa aesar
40274KT
1 ea. Each for $132.07

AA40274KU

 
alfa aesar
40274KU
5 ea. Each for $594.16
Description & Specifications

Specifications

Physical Form Plate
Quantity 1 ea.
Color White
Chemical Name or Material Magnesium Oxide Substrate
Odor Odorless
Name Note Polished Two Sides
Molecular Formula MgO
MDL Number MFCD00011109