Please login to your online account to display your discounted pricing

Magnesium Oxide (Powder/Certified ACS), Fisher Chemical


Quantity 500g
Packaging Poly Bottle

Manufacturer: fisher scientific company  M51500

 View more versions of this product

Catalog No. M51-500

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Chemical Identifiers

Assay ≥95.0%
Physical Form Powder/Solid
Boiling Point 3600°C
Purity Grade Certified ACS Grade
Formula Weight 40.3
Melting Point 2800°C
Quantity 500g
Vapor Pressure 0 hPa at 20°C
Viscosity [NULL]
Color White
pH 10.3
Density 3.580g/cm³
Heavy Metals (as Pb) 0.003% max.
Loss on Ignition 2.0% max.
Manganese (Mn) 5ppm max.
Nitrate 0.005% max.
CAS 1309-48-4
ChemAlert Storage Symbol Gray
Synonym Magnesia
Calcium (Ca) 0.05% max.
Chloride 0.01% max.
Chemical Name or Material Magnesium Oxide
Identification Pass Test
Packaging Poly Bottle
Potassium (K) 0.005% max.
Sodium (Na) 0.5% max.
Molecular Formula MgO
MDL Number MFCD00011109
Water Soluble Substances 0.4% max.
Barium (Ba) 0.005% max.
Insoluble in Hydrochloric Acid 0.02% max.
Iron (Fe) 0.01% max.
Assay Percent Range ≥95.0 %
Strontium (Sr) 0.005% max.
Sulfate and Sulfide 0.02% max.

Specifications

Chemical Name or Material Magnesium Oxide
CAS 1309-48-4
Purity Grade Certified ACS Grade
Physical Form Powder/Solid
Assay ≥95.0%
Assay Percent Range ≥95.0 %
Molecular Formula MgO
Formula Weight 40.3
MDL Number MFCD00011109
Synonym Magnesia
Accessories
 

SDS & Safety Documents