Please login to your online account to display your discounted pricing

Magnesium Oxide (Powder/Certified ACS), Fisher Chemical

$637.00 - $2,848.92

Chemical Identifiers

Chemical Name or Material Magnesium Oxide
CAS 1309-48-4
Purity Grade Certified ACS Grade
Physical Form Powder/Solid
Assay ≥95.0%
Assay Percent Range ≥95.0 %
Molecular Formula MgO
Formula Weight 40.3
MDL Number MFCD00011109
Synonym Magnesia
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

M51-100

  SDS
fisher scientific company
M51100
Glass Bottle 100g Each for $637.00
Case of 6 Each for $3,822.00

M51-500

  SDS
fisher scientific company
M51500
Poly Bottle 500g Each for $2,848.92
Case of 6 Each for $17,093.52
Description & Specifications

Specifications

Assay ≥95.0%
Purity Grade Certified ACS Grade
Physical Form Powder/Solid
Formula Weight 40.3
Boiling Point 3600°C
Melting Point 2800°C
Quantity 100g
Color White
pH 10.3
Density 3.580g/cm³
Vapor Pressure 0 hPa at 20°C
Viscosity [NULL]
Iron (Fe) 0.01% max.
Loss on Ignition 2.0% max.
Nitrate 0.005% max.
Potassium (K) 0.005% max.
Sodium (Na) 0.5% max.
MDL Number MFCD00011109
CAS 1309-48-4
ChemAlert Storage Symbol Gray
Synonym Magnesia
Calcium (Ca) 0.05% max.
Chloride 0.01% max.
Heavy Metals (as Pb) 0.003% max.
Packaging Glass Bottle
Strontium (Sr) 0.005% max.
Sulfate and Sulfide 0.02% max.
Water Soluble Substances 0.4% max.
Barium (Ba) 0.005% max.
Chemical Name or Material Magnesium Oxide
Identification Pass Test
Insoluble in Hydrochloric Acid 0.02% max.
Manganese (Mn) 5ppm max.
Assay Percent Range ≥95.0 %
Molecular Formula MgO
Safety and Handling