missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Magnesium oxide, >99% (metals basis), Alfa Aesar™

S.A. >7.3m2/g

Manufacturer:  Alfa Aesar 4473322

 View more versions of this product

Catalog No. AA4473322



Specifications

Specifications

Magnesium oxide
>99% (Metal basis)
MFCD00011109
magnesium oxide, magnesia, seawater magnesia, magnesium oxide, causmag, granmag, maglite, periclase, seasorb, animag
O=[Mg]
40.304
40.3
White
1309-48-4
MgO
White powder
CPLXHLVBOLITMK-UHFFFAOYSA-N
oxomagnesium
14792
Powder
100g

Chemical Identifiers

1309-48-4
40.304
CPLXHLVBOLITMK-UHFFFAOYSA-N
14792
O=[Mg]
MgO
MFCD00011109
magnesium oxide, magnesia, seawater magnesia, magnesium oxide, causmag, granmag, maglite, periclase, seasorb, animag
oxomagnesium
Safety and Handling

Safety and Handling

EINECSNumber : 215-171-9

SDS Documents