Please login to your online account to display your discounted pricing

Magnesium oxide, 99.998% (metals basis), Alfa Aesar™, Puratronic™

$123.41 - $1,779.23

Chemical Identifiers

Chemical Name or Material Magnesium oxide
CAS 1309-48-4
Physical Form Powder
Assay Percent Range 99.998% (Metal basis)
Molecular Formula MgO
Formula Weight 40.3
MDL Number MFCD00011109
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1080030

  SDS
alfa aesar
1080030
250g Each for $1,779.23

AA1080009

  SDS
alfa aesar
1080009
10g Each for $123.41

AA1080018

  SDS
alfa aesar
1080018
50g Each for $470.11
Description & Specifications

Specifications

Chemical Name or Material Magnesium oxide
CAS 1309-48-4
EINECS Number 215-171-9
Physical Form Powder
Assay Percent Range 99.998% (Metal basis)
Molecular Formula MgO
Appearance White powder
Formula Weight 40.3
MDL Number MFCD00011109
Color White
Quantity 250g