missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Magnesium oxide, 99.998% (metals basis), Alfa Aesar™, Puratronic™

Manufacturer:  Thermo Fisher Scientific 1080030

 View more versions of this product

Catalog No. AA1080030



Specifications

Specifications

Magnesium oxide
99.998% (Metal basis)
MFCD00011109
animag
O=[Mg]
40.304
40.3
White
1309-48-4
MgO
White powder
CPLXHLVBOLITMK-UHFFFAOYSA-N
oxomagnesium
14792
Powder
250g

Chemical Identifiers

1309-48-4
40.304
CPLXHLVBOLITMK-UHFFFAOYSA-N
14792
O=[Mg]
MgO
MFCD00011109
animag
oxomagnesium
Safety and Handling

Safety and Handling

EINECSNumber : 215-171-9

Documents