missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Magnesium oxide, 99.99%, (trace metal basis), extra pure, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 193430100
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysisSpecifications
3600.0°C | |
2800.0°C | |
1309-48-4 | |
Powder | |
99.99% | |
MgO | |
01,633; 02,256; 07,221; 16,200 | |
magnesium oxide, magnesia, seawater magnesia, causmag, granmag, maglite, periclase, seasorb, animag | |
CPLXHLVBOLITMK-UHFFFAOYSA-N | |
oxomagnesium | |
14792 | |
99.99% | |
Glass bottle | |
3.5800g/mL |
Magnesium oxide | |
Extra Pure | |
White | |
10 g | |
MgO | |
MFCD00011109 | |
15, 5741 | |
Solubility in water: 6.2 mg/l (20°C), reacts generates heat and mg(oh)2. Other solubilities: soluble in diluted acids, insoluble in alcohol | |
O=[Mg] | |
40.30 | |
40.29 | |
Trace Metal Basis | |
3.58 |
Chemical Identifiers
1309-48-4 | |
40.30 | |
CPLXHLVBOLITMK-UHFFFAOYSA-N | |
14792 | |
O=[Mg] |
MgO | |
MFCD00011109 | |
magnesium oxide, magnesia, seawater magnesia, causmag, granmag, maglite, periclase, seasorb, animag | |
oxomagnesium |
Safety and Handling
EINECSNumber : 215-171-9
RUO – Research Use Only