missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Magnesium oxide, 96%, Light, Alfa Aesar™

Bulk density: 4-8 lb./cu.ft.

Manufacturer:  Alfa Aesar 12287A1

 View more versions of this product

Catalog No. AA12287A1



Specifications

Specifications

Magnesium Oxide, Light
96%
MFCD00011109
magnesium oxide, magnesia, seawater magnesia, magnesium oxide, causmag, granmag, maglite, periclase, seasorb, animag
O=[Mg]
40.304
40.3
3.58g/cm3 at 20°C
3600°C
2852°C
1309-48-4
MgO
White powder
CPLXHLVBOLITMK-UHFFFAOYSA-N
oxomagnesium
14792
Powder
−325
White
1kg

Chemical Identifiers

1309-48-4
40.304
CPLXHLVBOLITMK-UHFFFAOYSA-N
14792
O=[Mg]
MgO
MFCD00011109
magnesium oxide, magnesia, seawater magnesia, magnesium oxide, causmag, granmag, maglite, periclase, seasorb, animag
oxomagnesium
Safety and Handling

Safety and Handling

EINECSNumber : 215-171-9

SDS Documents