missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Magnesium oxide, 96%, Light, Alfa Aesar™

Bulk density: 4-8 lb./cu.ft.

Manufacturer:  Thermo Fisher Scientific 1228722

 View more versions of this product

Catalog No. AA1228722



Specifications

Specifications

Magnesium oxide, light
96%
MFCD00011109
animag
CPLXHLVBOLITMK-UHFFFAOYSA-N
oxomagnesium
14792
-325 Mesh Powder
1.736
White
100g
1309-48-4
MgO
White powder
Soluble in acid and ammonia. Insoluble in water and ethanol.
O=[Mg]
40.304
40.3
3.58
3600°C
2852°C

Chemical Identifiers

1309-48-4
40.304
CPLXHLVBOLITMK-UHFFFAOYSA-N
14792
O=[Mg]
MgO
MFCD00011109
animag
oxomagnesium
Safety and Handling

Safety and Handling

EINECSNumber : 215-171-9

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents