missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Magnesium oxide, 95% min, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 043195A3

 View more versions of this product

Catalog No. AA43195A3



Description

Description

For plasma spraying, Refractory applications, electrical insulation, pharmaceuticals, cosmetics, paper manufacture, adsorption and catalysis, and in semiconductors, reflector in optical instruments. Magnesium oxide is used as a laboratory reagent and an insulator in industrial cables. It acts as a reference material for calorimetry. It is widely used as a basic refractory material for crucibles, a protective coating in plasma displays, an oxide barrier in spin-tunneling devices, an optical material, a component of tubular construction in heating elements, and also in libraries for preserving books. Magnesium oxide microspheres used in enrichment method of polycyclic aromatic hydrocarbons.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • For plasma spraying, Refractory applications, electrical insulation, pharmaceuticals, cosmetics, paper manufacture, adsorption and catalysis, and in semiconductors, reflector in optical instruments
Specifications

Specifications

3600°C
2852°C
Powder
95% min
MFCD00011109
magnesium oxide, magnesia, seawater magnesia, causmag, granmag, maglite, periclase, seasorb, animag
CPLXHLVBOLITMK-UHFFFAOYSA-N
oxomagnesium
14792
≥95%
1.736
Magnesium oxide
1309-48-4
2 kg
MgO
14,5677
Slightly soluble in water,(solubility increased by CO2); Soluble in acids and ammonium salt solution; Insoluble in alcoholSoluble in acid and ammonia; Insoluble in water and ethanol.
O=[Mg]
40.30
40.3
Odorless
3.58 g/mL

Chemical Identifiers

1309-48-4
40.30
CPLXHLVBOLITMK-UHFFFAOYSA-N
14792
O=[Mg]
MgO
MFCD00011109
magnesium oxide, magnesia, seawater magnesia, causmag, granmag, maglite, periclase, seasorb, animag
oxomagnesium
Safety and Handling

Safety and Handling

EINECSNumber : 215-171-9

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only