Please login to your online account to display your discounted pricing

Magnesium Oxide, ≥95%, (ACS Reagent Grade), MP Biomedicals


Quantity 500g

Manufacturer: mp biomedicals  0219488390

 View more versions of this product

Catalog No. ICN19488390

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 500g
Chemical Name or Material Magnesium Oxide
CAS 1309-48-4
Purity Grade ACS Reagent
Assay Percent Range ≥95%
Molecular Formula MgO
Formula Weight 40.3g/mol
Recommended Storage Room Temperature

Specifications

Chemical Name or Material Magnesium Oxide
CAS 1309-48-4
Purity Grade ACS Reagent
Assay Percent Range ≥95%
Molecular Formula MgO
Formula Weight 40.3g/mol
Accessories

Room Temperature