Please login to your online account to display your discounted pricing

Magnesium Oxide, ≥95%, (ACS Reagent Grade), MP Biomedicals


Quantity 100g

Manufacturer: mp biomedicals  19488380

 View more versions of this product

Catalog No. ICN19488380

  • / Each
 

Description & Specifications

Chemical Identifiers

Purity Grade ACS Reagent Grade
Formula Weight 40.3
Quantity 100g
Chemical Name or Material Magnesium Oxide
Molecular Formula MgO
CAS 1309-48-4
Assay Percent Range ≥95%
Recommended Storage Room Temperature

Specifications

Chemical Name or Material Magnesium Oxide
CAS 1309-48-4
Purity Grade ACS Reagent Grade
Assay Percent Range ≥95%
Molecular Formula MgO
Formula Weight 40.3
Accessories

Room Temperature