missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Magnesium Oxide, ≥95%, (ACS Reagent Grade), MP Biomedicals

$137.04 - $137.04

Chemical Identifiers

CAS 1309-48-4
Assay Percent Range ≥95%
Molecular Formula MgO
Formula Weight 40.3
Chemical Name or Material Magnesium Oxide
Grade ACS Reagent Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ICN19488380 Mp Biomedicals
19488380
100g Each for $137.04
Specifications Description & Specifications

Chemical Identifiers

1309-48-4
MgO
Magnesium Oxide
≥95%
40.3
ACS Reagent Grade

Specifications

1309-48-4
MgO
100g
ACS Reagent Grade
≥95%
40.3
Magnesium Oxide