Please login to your online account to display your discounted pricing

Magnesium oxide, 2 part ceramic adhesive paste

Chemical Identifiers

Chemical Name or Material Magnesium oxide, 2 part ceramic adhesive paste
Name Note Mix ratio (w/w): 1.5:1 powder to liquid. Cure conditions: 1-4 hours at room temperature, 2 hours at 94°C
MDL Number MFCD00011109
View More Specs
Products
Catalog Number Mfr. No. Price Quantity    

AA4313536

 
alfa aesar
4313536
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA43135A4

 
alfa aesar
43135A4
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 500g
Health Hazard 1 H314-H335-H303: Causes severe skin burns and eye damage. May cause respiratory irritation. May be harmful if swallowed.
Chemical Name or Material Magnesium oxide, 2 part ceramic adhesive paste
Name Note Mix ratio (w/w): 1.5:1 powder to liquid. Cure conditions: 1-4 hours at room temperature, 2 hours at 94°C
MDL Number MFCD00011109
Safety and Handling

H314-H335-H303: Causes severe skin burns and eye damage. May cause respiratory irritation. May be harmful if swallowed.