Please login to your online account to display your discounted pricing

Alfa Aesar™ Magnesium oxalate dihydrate, Puratronic™, 99.999% (metals basis)

$57.53 - $57.53

Chemical Identifiers

Chemical Name or Material Magnesium oxalate dihydrate
CAS 547-66-0
Purity Grade Puratronic™
Formula Weight 148.36g/mol (112.32g/mol Anhydrous)
MDL Number MFCD00287279
UN Number UN3288
Merck Index 14,5676
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1299304

 
alfa aesar™
12993-04
2g Each for $57.53
Description & Specifications

Specifications

Odor Odorless
Quantity 2g
Melting Point 150°C (dec.)
Quantity 2g
Chemical Name or Material Magnesium oxalate dihydrate
CAS 547-66-0
Purity Grade Puratronic™
Physical Form Powder
Assay 99.999%
Assay Percent Notes (metals basis)
Formula Weight 148.36g/mol (112.32g/mol Anhydrous)
MDL Number MFCD00287279
UN Number UN3288
Merck Index 14,5676
Solubility Information Soluble in dilute mineral acid; Practically insoluble in water; Insoluble in alcohol
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.
Health Hazard 3 GHS P Statement
P280-P301+P312-P312-P363-P322-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash contaminated clothing before reuse.
Specific measures (see label).
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
TSCA TSCA
Safety and Handling

Warning

GHS P Statement
P280-P301+P312-P312-P363-P322-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash contaminated clothing before reuse.
Specific measures (see label).
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.