Learn More
Alfa Aesar™ Magnesium oxalate dihydrate, 98.5+%
Manufacturer: Alfa Aesar™ 43976A3
Specifications
Magnesium oxalate dihydrate | |
C2H2MgO4+2 | |
UN3288 | |
magnesium oxalate, magnesium oxalate 1:1, magnesium oxalate mgc2o4, oxalic acid, magnesium salt 1:1, ethanedioic acid, magnesium salt 1:1 | |
C(=O)(C(=O)O)O.[Mg+2] | |
114.339 | |
148.36 (112.32 Anhydrous) | |
≥98.5% | |
150°C (decomposition) |
547-66-0 | |
MFCD00287279 | |
14,5676 | |
UHNWOJJPXCYKCG-UHFFFAOYSA-N | |
magnesium;oxalic acid | |
54611841 | |
Powder/Pieces | |
Odorless | |
2kg |
Chemical Identifiers
547-66-0 | |
114.339 | |
UHNWOJJPXCYKCG-UHFFFAOYSA-N | |
54611841 | |
C(=O)(C(=O)O)O.[Mg+2] |
C2H2MgO4+2 | |
MFCD00287279 | |
magnesium oxalate, magnesium oxalate 1:1, magnesium oxalate mgc2o4, oxalic acid, magnesium salt 1:1, ethanedioic acid, magnesium salt 1:1 | |
magnesium;oxalic acid |
Safety and Handling
GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.
GHS P Statement
P280-P301+P312-P312-P363-P322-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash contaminated clothing before reuse.
Specific measures (see label).
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
TSCA : TSCA