missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Magnesium oxalate dihydrate, 98.5+%

Manufacturer:  Alfa Aesar™ 4397636

 View more versions of this product

Catalog No. AA4397636



Specifications

Specifications

Magnesium oxalate dihydrate
C2H2MgO4+2
UN3288
magnesium oxalate, magnesium oxalate 1:1, magnesium oxalate mgc2o4, oxalic acid, magnesium salt 1:1, ethanedioic acid, magnesium salt 1:1
C(=O)(C(=O)O)O.[Mg+2]
114.339
148.36 (112.32 Anhydrous)
≥98.5%
150°C (decomposition)
547-66-0
MFCD00287279
14,5676
UHNWOJJPXCYKCG-UHFFFAOYSA-N
magnesium;oxalic acid
54611841
Powder/Pieces
Odorless
500g

Chemical Identifiers

547-66-0
114.339
UHNWOJJPXCYKCG-UHFFFAOYSA-N
54611841
C(=O)(C(=O)O)O.[Mg+2]
C2H2MgO4+2
MFCD00287279
magnesium oxalate, magnesium oxalate 1:1, magnesium oxalate mgc2o4, oxalic acid, magnesium salt 1:1, ethanedioic acid, magnesium salt 1:1
magnesium;oxalic acid
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
P280-P301+P312-P312-P363-P322-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash contaminated clothing before reuse.
Specific measures (see label).
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.

TSCA : TSCA

Documents