Please login to your online account to display your discounted pricing

Alfa Aesar™ Magnesium, Oil based standard solution, Specpure™, Mg 1000μg/g

Chemical Identifiers

Chemical Name or Material Magnesium Oil based standard solution
Name Note Mg 1000μg/g
Purity Grade Specpure™
MDL Number MFCD00085308
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1421918

  SDS
alfa aesar™
1421918
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 50g
MDL Number MFCD00085308
Physical Form Liquid
Name Note Mg 1000μg/g
Chemical Name or Material Magnesium Oil based standard solution
Purity Grade Specpure™