Please login to your online account to display your discounted pricing

Magnesium Nitrate Hexahydrate (Cryst./Certified ACS), Fisher Chemical


Quantity 2.5kg
Packaging Glass Bottle

Manufacturer: fisher scientific company  FLM46212

 View more versions of this product

Catalog No. M46-212

  • / Each
  • / Case of 4 Each
 

Description & Specifications

Chemical Identifiers

Chemical Name or Material Magnesium Nitrate Hexahydrate
CAS 13446-18-9
ChemAlert Storage Symbol Yellow
Health Hazard 2 WARNING!
Health Hazard 3 Emergency Overview
Irritating to eyes, respiratory system and skin. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Obtain medical attention. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .

NFPA

Health:2
Flammability:1
Instability:0
DOT Information DOT Class 5.1, : Oxidizer
Purity Grade Certified ACS Grade
Physical Form Solid
Assay 98 to 102%
Assay Percent Range 98 to 102 %
Molecular Formula MgN2O6 · 6 H2O
Ammonium 0.003% max.
Linear Formula Mg(NO3)2·6H2O
Formula Weight 256.41
MDL Number MFCD00149779
UN Number 1474
Barium (Ba) 0.005% max.
Calcium (Ca) 0.01% max.
Chloride 0.001% max.
Heavy Metals (as Pb) 5ppm max.
Identification Pass Test
Insoluble Matter 0.005% max.
Iron (Fe) 5ppm max.
Manganese (Mn) 5ppm max.
Packaging Glass Bottle
Phosphate 5ppm max.
Potassium (K) 0.005% max.
Sodium (Na) 0.005% max.
Strontium (Sr) 0.005% max.
Sulfate 0.005% max.
Boiling Point 330°C
Color White
Melting Point 89°C
pH 5.0 to 8.2
Quantity 2.5kg

Specifications

Chemical Name or Material Magnesium Nitrate Hexahydrate
CAS 13446-18-9
Purity Grade Certified ACS Grade
Physical Form Solid
Assay 98 to 102%
Assay Percent Range 98 to 102 %
Molecular Formula MgN2O6 · 6 H2O
Linear Formula Mg(NO3)2·6H2O
Formula Weight 256.41
MDL Number MFCD00149779
Accessories
 

Emergency Overview
Irritating to eyes, respiratory system and skin. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Obtain medical attention. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .

NFPA

Health:2
Flammability:1
Instability:0

WARNING!

SDS & Safety Documents