Magnesium nitrate hexahydrate, ACS, 98.0-102.0%, Alfa Aesar
As fertilizer, in manufacture of ammonium nitrate
Manufacturer:  Alfa Aesar 1156418
 View more versions of this product
Catalog No. AA1156418
Specifications
Specifications
Magnesium nitrate
13446-18-9
H12MgN2O12
MFUVDXOKPBAHMC-UHFFFAOYSA-N
magnesium;dinitrate;hexahydrate
202877
Crystalline
1.46
hexahydrate
98.0-102.0%
dusicnan horecnaty
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.[Mg+2]
256.403
256.41 (148.32 Anhydrous)
ACS
50g
Chemical Identifiers
13446-18-9
256.403
dusicnan horecnaty
magnesium;dinitrate;hexahydrate
H12MgN2O12
MFUVDXOKPBAHMC-UHFFFAOYSA-N
202877
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.[Mg+2]
Safety and Handling
Safety and Handling
storageNote1 : Hygroscopic
Documents