missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Magnesium nitrate hexahydrate, extra pure, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 447540050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
330.0°C | |
89.0°C | |
5 kg | |
Mg(NO3)2·6H2O | |
93.3°C | |
magnesium nitrate hexahydrate, magnesium dinitrate hexahydrate, unii-v85k20ljmk, dusicnan horecnaty czech, v85k20ljmk, nitric acid, magnesium salt, hexahydrate, magnesium ii nitrate 1:2 , hexahydrate, magnesium 2+ ion hexahydrate dinitrate, magnesium, reference standard solution, dusicnan horecnaty | |
MFUVDXOKPBAHMC-UHFFFAOYSA-N | |
magnesium;dinitrate;hexahydrate | |
202877 | |
Extra Pure |
Magnesium Nitrate Hexahydrate | |
13446-18-9 | |
H12MgN2O12 | |
MFCD00149779 | |
15,5738 | |
Solubility in water: 420g/L (20°C). Other solubilities: freely soluble in alcohol, liquid nh3 | |
O.O.O.O.O.O.[Mg++].[O-][N+]([O-])=O.[O-][N+]([O-])=O | |
256.40 | |
256.4 | |
Plastic Drum |
Chemical Identifiers
13446-18-9 | |
256.40 | |
MFUVDXOKPBAHMC-UHFFFAOYSA-N | |
202877 | |
O.O.O.O.O.O.[Mg++].[O-][N+]([O-])=O.[O-][N+]([O-])=O |
H12MgN2O12 | |
MFCD00149779 | |
magnesium nitrate hexahydrate, magnesium dinitrate hexahydrate, unii-v85k20ljmk, dusicnan horecnaty czech, v85k20ljmk, nitric acid, magnesium salt, hexahydrate, magnesium ii nitrate 1:2 , hexahydrate, magnesium 2+ ion hexahydrate dinitrate, magnesium, reference standard solution, dusicnan horecnaty | |
magnesium;dinitrate;hexahydrate |
RUO – Research Use Only