missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Magnesium nitrate hexahydrate, 99.97% (metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 8760930

 View more versions of this product

Catalog No. AA8760930



Specifications

Specifications

Magnesium nitrate hexahydrate
99.97% (metals basis)
MFCD00149779
MFUVDXOKPBAHMC-UHFFFAOYSA-N
magnesium;dinitrate;hexahydrate
202877
Crystalline
250g
13446-18-9
H12MgN2O12
magnesium oil based standard solution specpure mg 5000g/g, magnesium oil based standard solution specpure mg 1000g/g, magnesium plasma standard solution specpure mg 10000g/ml, magnesium plasma standard solution specpure mg 1000g/ml, magnesium aas standard solution specpure mg 1000g/ml, magnesium nitrate hexahydrate 250g, nitric acid magnesium salt hexahydrate, magnesium ii nitrate 1:2 hexahydrate, magnesium nitrate hydrate puratronic r, magnesium 2+ ion hexahydrate dinitrate
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.[Mg+2]
256.403
256.41 (148.32 anhydrous)
1.46 g/mL

Chemical Identifiers

13446-18-9
256.403
MFUVDXOKPBAHMC-UHFFFAOYSA-N
202877
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.[Mg+2]
H12MgN2O12
MFCD00149779
magnesium oil based standard solution specpure mg 5000g/g, magnesium oil based standard solution specpure mg 1000g/g, magnesium plasma standard solution specpure mg 10000g/ml, magnesium plasma standard solution specpure mg 1000g/ml, magnesium aas standard solution specpure mg 1000g/ml, magnesium nitrate hexahydrate 250g, nitric acid magnesium salt hexahydrate, magnesium ii nitrate 1:2 hexahydrate, magnesium nitrate hydrate puratronic r, magnesium 2+ ion hexahydrate dinitrate
magnesium;dinitrate;hexahydrate
Safety and Handling

Safety and Handling

EINECSNumber : 233-826-7

SDS Documents