Please login to your online account to display your discounted pricing

Alfa Aesar™ Magnesium methoxide, 7-8% in methanol

MgO films

Chemical Identifiers

Chemical Name or Material Magnesium methoxide
CAS 109-88-6
Formula Weight 86.38
MDL Number MFCD00053671
UN Number UN1230
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4044830

  SDS
alfa aesar™
4044830
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA40448A1

  SDS
alfa aesar™
40448A1
1kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA40448A3

  SDS
alfa aesar™
40448A3
2kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 86.38
Refractive Index 1.3380
Odor Alcohol-like
Sensitivity Moisture Sensitive
Color Colorless to pale yellow
TSCA TSCA
MDL Number MFCD00053671
Quantity 250g
Health Hazard 2 GHS H Statement
H225-H311-H331-H370-H302
Highly flammable liquid and vapor.
Toxic in contact with skin.
Toxic if inhaled.
Causes damage to organs.
Harmful if swallowed.
CAS 109-88-6
Concentration 7 to 8% in methanol
Chemical Name or Material Magnesium methoxide
Flash Point 14° (57°F)
Physical Form Liquid
Health Hazard 3 GHS P Statement
P210-P260-P303+P361+P353-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
UN Number UN1230
Health Hazard 1 Danger
Density 0.815g/mL
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: METHANOL
Safety and Handling

Danger

GHS P Statement
P210-P260-P303+P361+P353-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H311-H331-H370-H302
Highly flammable liquid and vapor.
Toxic in contact with skin.
Toxic if inhaled.
Causes damage to organs.
Harmful if swallowed.