missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Magnesium iodide, ultra dry, 99.996% (metals basis), Alfa Aesar™

Light Sensitive and Hygroscopic

Manufacturer:  Thermo Fisher Scientific 4431014

 View more versions of this product

Catalog No. AA4431014



Specifications

Specifications

Magnesium iodide, ultra dry
99.996% (Metal basis)
MFCD00049485
Soluble in ether,alcohol and ammonia.
Powder
Odorless
White
25g
10377-58-9
I2Mg
White powder
278.12
4.43g/cm3 at 20°C
Hygroscopic; Light sensitive
637°C (decomposition)

Chemical Identifiers

10377-58-9
I2Mg
MFCD00049485
Safety and Handling

Safety and Handling

EINECSNumber : 233-825-1

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents