Please login to your online account to display your discounted pricing

Magnesium iodide, ultra dry, 99.996% (metals basis), Alfa Aesar™

Light Sensitive and Hygroscopic

$483.34 - $483.34

Chemical Identifiers

Chemical Name or Material Magnesium iodide, ultra dry
CAS 10377-58-9
Physical Form Powder
Assay Percent Range 99.996% (Metal basis)
Molecular Formula I2Mg
Formula Weight 278.12
MDL Number MFCD00049485
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4431014

  SDS
alfa aesar
4431014
25g Each for $483.34

AA4431022

  SDS
alfa aesar
4431022
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Powder
Formula Weight 278.12
Quantity 25g
Color White
Density 4.43g/cm3 at 20°C
Chemical Name or Material Magnesium iodide, ultra dry
EINECS Number 233-825-1
Odor Odorless
Assay Percent Range 99.996% (Metal basis)
Molecular Formula I2Mg
Appearance White powder
MDL Number MFCD00049485
CAS 10377-58-9