missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Magnesium ICP Standard, 1000 ppm Mg in 3% HNO3, Ricca Chemical
$265.44 - $551.34
Chemical Identifiers
CAS | 1309-48-4 |
---|---|
Molecular Formula | MgO |
Molecular Weight (g/mol) | 40.30 |
MDL Number | MFCD00011109 |
InChI Key | CPLXHLVBOLITMK-UHFFFAOYSA-N |
PubChem CID | 14792 |
IUPAC Name | oxomagnesium |
SMILES | O=[Mg] |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
PMG1KN-100
|
Ricca Chemical Company
PMG1KN100 |
100 mL | Natural LDPE Bottle |
Each for $265.44
|
|
||||
PMG1KN-500
|
Ricca Chemical Company
PMG1KN500 |
500 mL | Natural Poly Bottle |
Each for $551.34
|
|
||||
Chemical Identifiers
1309-48-4 | |
40.30 | |
CPLXHLVBOLITMK-UHFFFAOYSA-N | |
oxomagnesium |
MgO | |
MFCD00011109 | |
14792 | |
O=[Mg] |
Specifications
1309-48-4,7697-37-2,7732-18-5 | |
100 mL | |
97.0,5.12,0.16 | |
1,000ppm Mg | |
MFCD00011109 | |
O=[Mg] | |
40.30 | |
1 mL = 1 mg Mg | |
Magnesium ICP Standard |
Natural LDPE Bottle | |
93.0,4.92,0.15 | |
Colorless | |
MgO | |
CPLXHLVBOLITMK-UHFFFAOYSA-N | |
oxomagnesium | |
14792 | |
Liquid |