Please login to your online account to display your discounted pricing

Magnesium ICP Standard, 1mL = 10mg Mg (10,000ppm Mg), Ricca Chemical


Quantity 50mL
Packaging Poly natural

Manufacturer: ricca chemical company  RIPMG10KN50

 View more versions of this product

Catalog No. PMG10KN-50

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Liquid
Color Clear colorless
Quantity 50mL
Equivalence 1 mL = 10 mg Mg
Concentration or Composition Notes 10,000ppm Mg
CAS Max % 87.0
CAS 7697-37-2
CAS Min % 84.0
CAS Max % 1.45
CAS Min % 12.84
CAS Min % 1.39
CAS Max % 13.36
Packaging Poly natural
CAS 1309-48-4
Chemical Name or Material Magnesium ICP Standard
CAS 7732-18-5

Specifications

Chemical Name or Material Magnesium ICP Standard
CAS 7732-18-5
CAS Min % 84.0
CAS Max % 87.0
CAS 7697-37-2
CAS Min % 12.84
CAS Max % 13.36
CAS 1309-48-4
CAS Min % 1.39
CAS Max % 1.45
Physical Form Liquid