missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Magnesium ICP Standard, 10,000 ppm Mg in 5% HNO3, Ricca Chemical
Supplier: Ricca Chemical Company PMG10KN500
Specifications
Magnesium ICP Standard | |
Natural Poly Bottle | |
84.0,12.84,1.39 | |
Colorless | |
MgO | |
CPLXHLVBOLITMK-UHFFFAOYSA-N | |
oxomagnesium | |
14792 | |
Liquid |
1309-48-4,7697-37-2,7732-18-5 | |
500 mL | |
87.0,13.36,1.45 | |
10,000ppm Mg | |
MFCD00011109 | |
O=[Mg] | |
40.30 | |
1 mL = 10 mg Mg |
Chemical Identifiers
1309-48-4 | |
40.30 | |
CPLXHLVBOLITMK-UHFFFAOYSA-N | |
oxomagnesium |
MgO | |
MFCD00011109 | |
14792 | |
O=[Mg] |