Please login to your online account to display your discounted pricing

Magnesium ICP Standard, 1mL = 10mg Mg (10,000ppm Mg), Ricca Chemical

$156.97 - $628.12

Chemical Identifiers

Chemical Name or Material Magnesium ICP Standard
CAS 7732-18-5
CAS Min % 84.0
CAS Max % 87.0
CAS 7697-37-2
CAS Min % 12.84
CAS Max % 13.36
CAS 1309-48-4
CAS Min % 1.39
CAS Max % 1.45
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

PMG10KN-50

 
ricca chemical company
RIPMG10KN50
Poly natural 50mL Each for $156.97

PMG10KN-100

 
ricca chemical company
RIPMG10KN100
Poly natural 100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

PMG10KN-500

 
ricca chemical company
RIPMG10KN500
Poly natural 500mL Each for $628.12
Description & Specifications

Specifications

Physical Form Liquid
Color Clear colorless
Quantity 50mL
Equivalence 1 mL = 10 mg Mg
Concentration or Composition Notes 10,000ppm Mg
CAS Max % 87.0
CAS 7697-37-2
CAS Min % 84.0
CAS Max % 1.45
CAS Min % 12.84
CAS Min % 1.39
CAS Max % 13.36
Packaging Poly natural
CAS 1309-48-4
Chemical Name or Material Magnesium ICP Standard
CAS 7732-18-5