Learn More
Magnesium hydroxide, 95%, pure, Thermo Scientific Chemicals
$61.18 - $110.79
Chemical Identifiers
CAS | 1309-42-8 |
---|---|
Molecular Formula | H2MgO2 |
Molecular Weight (g/mol) | 58.32 |
MDL Number | MFCD00011104 |
InChI Key | VTHJTEIRLNZDEV-UHFFFAOYSA-L |
Synonym | milk of magnesia, magnesium dihydroxide, brucite, magnesium hydroxide suspension, magnesium hydroxide gilumag, magnesiumhydroxid, magnesiumhydroxide, magnesia,, acmc-1bqga, magnesium 2+ hydroxide |
PubChem CID | 73981 |
ChEBI | CHEBI:6637 |
SMILES | [OH-].[OH-].[Mg++] |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC315720250
|
Thermo Scientific Chemicals
315720250 |
25 g | Plastic bottle |
Each for $61.18
|
|
||||
AC315725000
|
Thermo Scientific Chemicals
315725000 |
500 g | Plastic bottle |
Each for $110.79
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1309-42-8 | |
58.32 | |
VTHJTEIRLNZDEV-UHFFFAOYSA-L | |
73981 | |
[OH-].[OH-].[Mg++] |
H2MgO2 | |
MFCD00011104 | |
milk of magnesia, magnesium dihydroxide, brucite, magnesium hydroxide suspension, magnesium hydroxide gilumag, magnesiumhydroxid, magnesiumhydroxide, magnesia,, acmc-1bqga, magnesium 2+ hydroxide | |
CHEBI:6637 |
Specifications
350°C | |
White | |
25 g | |
H2MgO2 | |
MFCD00011104 | |
milk of magnesia, magnesium dihydroxide, brucite, magnesium hydroxide suspension, magnesium hydroxide gilumag, magnesiumhydroxid, magnesiumhydroxide, magnesia,, acmc-1bqga, magnesium 2+ hydroxide | |
VTHJTEIRLNZDEV-UHFFFAOYSA-L | |
magnesium;dihydroxide | |
73981 | |
58.33 | |
Pure | |
Magnesium hydroxide, 95% |
1309-42-8 | |
Powder | |
94% min. (Complexometry) | |
Mg(OH)2 | |
15, 5734 | |
Solubility in water: insoluble | |
[OH-].[OH-].[Mg++] | |
58.32 | |
CHEBI:6637 | |
95% | |
Plastic bottle |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
AIR SENSITIVE
EINECSNumber : 215-170-3
RTECSNumber : OM3570000
TSCA : TSCA
RUO – Research Use Only