missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Magnesium hydroxide, 95 to 100.5%, Alfa Aesar™

Flame Retardant in Plastics, Such As PVC, As It Produces Less Toxic Fumes

Manufacturer:  Alfa Aesar 12367A4

 View more versions of this product

Catalog No. AA12367A4



Specifications

Specifications

Magnesium hydroxide
95-100.5%
MFCD00011104
milk of magnesia, magnesium dihydroxide, brucite, magnesium hydroxide suspension, magnesium hydroxide gilumag, magnesiumhydroxid, magnesiumhydroxide, magnesia,, acmc-1bqga, magnesium 2+ hydroxide
[OH-].[OH-].[Mg+2]
58.319
CHEBI:6637
Powder
White
1309-42-8
H2MgO2
White powder
VTHJTEIRLNZDEV-UHFFFAOYSA-L
magnesium;dihydroxide
73981
58.33
Odorless
2.5kg

Chemical Identifiers

1309-42-8
58.319
VTHJTEIRLNZDEV-UHFFFAOYSA-L
73981
magnesium;dihydroxide
H2MgO2
MFCD00011104
milk of magnesia, magnesium dihydroxide, brucite, magnesium hydroxide suspension, magnesium hydroxide gilumag, magnesiumhydroxid, magnesiumhydroxide, magnesia,, acmc-1bqga, magnesium 2+ hydroxide
CHEBI:6637
[OH-].[OH-].[Mg+2]
Safety and Handling

Safety and Handling

H315-H319-H335: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

EINECSNumber : 215-170-3

Documents