missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Magnesium hydroxide, 95 to 100.5%, Alfa Aesar™
Flame Retardant in Plastics, Such As PVC, As It Produces Less Toxic Fumes
Manufacturer: Alfa Aesar 1236722
Specifications
Magnesium hydroxide | |
95-100.5% | |
MFCD00011104 | |
milk of magnesia, magnesium dihydroxide, brucite, magnesium hydroxide suspension, magnesium hydroxide gilumag, magnesiumhydroxid, magnesiumhydroxide, magnesia,, acmc-1bqga, magnesium 2+ hydroxide | |
[OH-].[OH-].[Mg+2] | |
58.319 | |
CHEBI:6637 | |
Powder | |
White |
1309-42-8 | |
H2MgO2 | |
White powder | |
VTHJTEIRLNZDEV-UHFFFAOYSA-L | |
magnesium;dihydroxide | |
73981 | |
58.33 | |
Odorless | |
100g |
Chemical Identifiers
1309-42-8 | |
58.319 | |
VTHJTEIRLNZDEV-UHFFFAOYSA-L | |
73981 | |
magnesium;dihydroxide |
H2MgO2 | |
MFCD00011104 | |
milk of magnesia, magnesium dihydroxide, brucite, magnesium hydroxide suspension, magnesium hydroxide gilumag, magnesiumhydroxid, magnesiumhydroxide, magnesia,, acmc-1bqga, magnesium 2+ hydroxide | |
CHEBI:6637 | |
[OH-].[OH-].[Mg+2] |
Safety and Handling
H315-H319-H335: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
EINECSNumber : 215-170-3