missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Magnesium hydrogen phosphate trihydrate, 98%, extra pure, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 264780050

 View more versions of this product

Catalog No. AC264780050



Specifications

Specifications

Magnesium hydrogen phosphate trihydrate
7782-75-4
HMgO43H2O
MFCD00150390
acmc-20aklg, dimagnesium phosphate, kc defense, magnesium 2+ ion trihydrate hydrogen phosphate, magnesium 2+ trihydrate hydrogen phosphate, magnesium dibasic phosphate trihydrate, magnesium hydrogen phosphate trihydrate, magnesium phosphate dibasic trihydrate, newberyite, unii-hf539g9l3q
Solubility in water: 0.25g/L (20°C). Other solubilities: soluble in diluted acids
O.O.O.OP(=O)([O-])[O-].[Mg+2]
174.34
174.34
98%
200ppm max
2.1300g/mL
50ppm max.
Solubility in water: 0.25g/L (20°C). Other solubilities: soluble in diluted acids
300ppm max.
5g
c.p., 98%
97.5% min. (Complexometry)
MgHPO4·3H2O
14, 5681
1ppm max.
OKIWLDVQGKRUNR-UHFFFAOYSA-L
magnesium;hydrogen phosphate;trihydrate
16204530
Powder
Extra Pure
200ppm max.
20ppm max.
Glass bottle
2.13
White

Chemical Identifiers

7782-75-4
174.34
OKIWLDVQGKRUNR-UHFFFAOYSA-L
16204530
O.O.O.OP(=O)([O-])[O-].[Mg+2]
HMgO43H2O
MFCD00150390
acmc-20aklg, dimagnesium phosphate, kc defense, magnesium 2+ ion trihydrate hydrogen phosphate, magnesium 2+ trihydrate hydrogen phosphate, magnesium dibasic phosphate trihydrate, magnesium hydrogen phosphate trihydrate, magnesium phosphate dibasic trihydrate, newberyite, unii-hf539g9l3q
magnesium;hydrogen phosphate;trihydrate
SDS Documents