missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Magnesium hydrogen phosphate trihydrate, 98%, extra pure, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 264780010
Specifications
Magnesium hydrogen phosphate trihydrate | |
7782-75-4 | |
HMgO4P·3H2O | |
MFCD00150390 | |
acmc-20aklg, dimagnesium phosphate, kc defense, magnesium 2+ ion trihydrate hydrogen phosphate, magnesium 2+ trihydrate hydrogen phosphate, magnesium dibasic phosphate trihydrate, magnesium hydrogen phosphate trihydrate, magnesium phosphate dibasic trihydrate, newberyite, unii-hf539g9l3q | |
Solubility in water: 0.25g/L (20°C). Other solubilities: soluble in diluted acids | |
O.O.O.OP(=O)([O-])[O-].[Mg+2] | |
174.34 | |
174.34 | |
98% | |
200ppm max | |
2.1300g/mL | |
50ppm max. | |
Solubility in water: 0.25g/L (20°C). Other solubilities: soluble in diluted acids | |
300ppm max. | |
1kg |
c.p., 98% | |
97.5% min. (Complexometry) | |
MgHPO4·3H2O | |
14, 5681 | |
1ppm max. | |
OKIWLDVQGKRUNR-UHFFFAOYSA-L | |
magnesium;hydrogen phosphate;trihydrate | |
16204530 | |
Powder | |
Extra Pure | |
200ppm max. | |
20ppm max. | |
Plastic bottle | |
2.13 | |
White |
Chemical Identifiers
7782-75-4 | |
174.34 | |
OKIWLDVQGKRUNR-UHFFFAOYSA-L | |
16204530 | |
O.O.O.OP(=O)([O-])[O-].[Mg+2] |
HMgO4P·3H2O | |
MFCD00150390 | |
acmc-20aklg, dimagnesium phosphate, kc defense, magnesium 2+ ion trihydrate hydrogen phosphate, magnesium 2+ trihydrate hydrogen phosphate, magnesium dibasic phosphate trihydrate, magnesium hydrogen phosphate trihydrate, magnesium phosphate dibasic trihydrate, newberyite, unii-hf539g9l3q | |
magnesium;hydrogen phosphate;trihydrate |