missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Magnesium hydrogen phosphate trihydrate, 98%, for biochemistry, ACROS Organics™

$79.02 - $79.02

Chemical Identifiers

CAS 7782-75-4
Molecular Formula H7MgO7P
Molecular Weight (g/mol) 174.33
MDL Number MFCD00150390
InChI Key OKIWLDVQGKRUNR-UHFFFAOYSA-L
Synonym acmc-20aklg, dimagnesium phosphate, kc defense, magnesium 2+ ion trihydrate hydrogen phosphate, magnesium 2+ trihydrate hydrogen phosphate, magnesium dibasic phosphate trihydrate, magnesium hydrogen phosphate trihydrate, magnesium phosphate dibasic trihydrate, newberyite, unii-hf539g9l3q
PubChem CID 16204530
IUPAC Name magnesium;hydrogen phosphate;trihydrate
SMILES O.O.O.[Mg++].OP([O-])([O-])=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC448182500
View Documents
Thermo Fisher Scientific
AC448182500
250g Plastic bottle
Each for $79.02
 
AC448180010
SDS View Documents
Thermo Fisher Scientific
AC448180010
1kg Plastic bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

7782-75-4
174.33
OKIWLDVQGKRUNR-UHFFFAOYSA-L
16204530
O.O.O.[Mg++].OP([O-])([O-])=O
H7MgO7P
MFCD00150390
acmc-20aklg, dimagnesium phosphate, kc defense, magnesium 2+ ion trihydrate hydrogen phosphate, magnesium 2+ trihydrate hydrogen phosphate, magnesium dibasic phosphate trihydrate, magnesium hydrogen phosphate trihydrate, magnesium phosphate dibasic trihydrate, newberyite, unii-hf539g9l3q
magnesium;hydrogen phosphate;trihydrate

Specifications

7782-75-4
H7MgO7P
MFCD00150390
Solubility in water: 0.25g/L (20°C). Other solubilities: soluble in diluted acids
O.O.O.[Mg++].OP([O-])([O-])=O
174.33
174.34
Biochemistry
2.1300g/mL
(0.1 M in 5 M HCl) Clear colorless
0.03% max
Magnesium hydrogen phosphate trihydrate
97.5% min. (Complexometry)
MgHPO4·3H2O
acmc-20aklg, dimagnesium phosphate, kc defense, magnesium 2+ ion trihydrate hydrogen phosphate, magnesium 2+ trihydrate hydrogen phosphate, magnesium dibasic phosphate trihydrate, magnesium hydrogen phosphate trihydrate, magnesium phosphate dibasic trihydrate, newberyite, unii-hf539g9l3q
OKIWLDVQGKRUNR-UHFFFAOYSA-L
magnesium;hydrogen phosphate;trihydrate
16204530
98%
0.01% max
Plastic bottle
2.13
250g
SDS
Product Certifications