Please login to your online account to display your discounted pricing

Magnesium hydride, 95%, ACROS Organics™

$307.66 - $307.66

Chemical Identifiers

Chemical Name or Material Magnesium hydride
Assay ≥94%
Assay Percent Range 95%
Molecular Formula H2Mg
Linear Formula MgH2
Formula Weight 26.32
MDL Number MFCD00011101
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC378450500

  SDS
thermo fisher scientific
AC378450500
Plastic bottle 50g Each for $307.66
Description & Specifications

Specifications

Physical Form Powder
Assay ≥94%
Formula Weight 26.32
Quantity 50g
Color Light gray
Merck Index 15, 5733
Health Hazard 3 GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Handle under inert gas. Protect from moisture.
Wear protective gloves/protective clothing/eye protection/face protection.
Brush off loose particles from skin.
Linear Formula MgH2
Assay Percent Range 95%
Health Hazard 1 Danger
MDL Number MFCD00011101
Health Hazard 2 GHS P Statement
Flammable solid.
In contact with water releases flammable gases which may ignite spontaneously.
Packaging Plastic bottle
Molecular Formula H2Mg
CAS 7693-27-8
Chemical Name or Material Magnesium hydride

Reduction reagent

Safety and Handling

Danger

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Handle under inert gas. Protect from moisture.
Wear protective gloves/protective clothing/eye protection/face protection.
Brush off loose particles from skin.

GHS P Statement
Flammable solid.
In contact with water releases flammable gases which may ignite spontaneously.