Please login to your online account to display your discounted pricing

Magnesium hydride, 95%, ACROS Organics™


Quantity 50g
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  AC378450500

 View more versions of this product

Catalog No. AC378450500

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Powder
Assay ≥94%
Formula Weight 26.32
Quantity 50g
Color Light gray
Merck Index 15, 5733
Health Hazard 3 GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Handle under inert gas. Protect from moisture.
Wear protective gloves/protective clothing/eye protection/face protection.
Brush off loose particles from skin.
Assay Percent Range 95%
Linear Formula MgH2
MDL Number MFCD00011101
Health Hazard 1 Danger
Health Hazard 2 GHS P Statement
Flammable solid.
In contact with water releases flammable gases which may ignite spontaneously.
Chemical Name or Material Magnesium hydride
Packaging Plastic bottle
Molecular Formula H2Mg
CAS 7693-27-8

Specifications

Chemical Name or Material Magnesium hydride
Assay ≥94%
Assay Percent Range 95%
Molecular Formula H2Mg
Linear Formula MgH2
Formula Weight 26.32
MDL Number MFCD00011101
Accessories

Danger

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Handle under inert gas. Protect from moisture.
Wear protective gloves/protective clothing/eye protection/face protection.
Brush off loose particles from skin.

GHS P Statement
Flammable solid.
In contact with water releases flammable gases which may ignite spontaneously.

SDS & Safety Documents