Please login to your online account to display your discounted pricing

Alfa Aesar™ Magnesium hexafluorosilicate hexahydrate, 98%

Chemical Identifiers

Chemical Name or Material Magnesium hexafluorosilicate hexahydrate
CAS 18972-56-0
Molecular Formula MgSiF6·6H2O
Formula Weight 274.47 (166.39 Anhydrous)
MDL Number MFCD00150250
UN Number UN2853
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA131890C

  SDS
alfa aesar™
A131890C
10000g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA131890E

  SDS
alfa aesar™
A131890E
2500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 274.47 (166.39 Anhydrous)
Chemical Name or Material Magnesium hexafluorosilicate hexahydrate
TSCA TSCA
Odor Odorless
Assay 98%
Molecular Formula MgSiF6·6H2O
MDL Number MFCD00150250
CAS 18972-56-0
UN Number UN2853
Quantity 10000g
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301
Toxic if swallowed.
Health Hazard 3 GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: MAGNESIUM FLUOROSILICATE
Safety and Handling

Danger

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301
Toxic if swallowed.