Please login to your online account to display your discounted pricing

Magnesium hexafluorosilicate hexahydrate 96%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Magnesium hexafluorosilicate hexahydrate
Assay Percent Range 96%
Molecular Formula F6MgSi·6H2O
Formula Weight 274.47
MDL Number MFCD00150250
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC384670250

  SDS
thermo fisher scientific
AC384670250
Plastic bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC384671000

  SDS
thermo fisher scientific
AC384671000
Plastic bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Powder
Formula Weight 274.47
Quantity 25g
Color White
Merck Index 15, 5732
Packaging Plastic bottle
Assay Percent Range 96%
Chemical Name or Material Magnesium hexafluorosilicate hexahydrate
CAS 18972-56-0
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Toxic if inhaled.
Causes serious eye irritation.
Toxic if swallowed.
Toxic in contact with skin.
MDL Number MFCD00150250
Elemental Analysis 95% min. (ex Fluor)
Molecular Formula F6MgSi·6H2O
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Toxic if inhaled.
Causes serious eye irritation.
Toxic if swallowed.
Toxic in contact with skin.