missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Magnesium germanide, 99.99% (metals basis)

Manufacturer:  Alfa Aesar™ 4551706

 View more versions of this product

Catalog No. AA4551706



Specifications

Specifications

Magnesium germanide
(metals basis)
Mg2Ge
dimagnesium germanide, magnesium germanide, germane-magnesium 1/2
[Mg].[Mg].[Ge]
121.24
121.22
99.99%
5g
1310-52-7
GeMg2
MFCD11044441
MBWJVOFAOUBYNU-UHFFFAOYSA-N
germanium;magnesium
76518831
Pieces
3 to 6mm

Chemical Identifiers

1310-52-7
121.24
MBWJVOFAOUBYNU-UHFFFAOYSA-N
76518831
[Mg].[Mg].[Ge]
GeMg2
MFCD11044441
dimagnesium germanide, magnesium germanide, germane-magnesium 1/2
germanium;magnesium
SDS Documents